Tisk…

4-(2-Methylbutan-2-yl)cyclohexan-1-amine hydrochloride 95%

BLDPBD00892311-1GEA 4940 CZK
BLDPBD00892311-1G BLDPBD00892311-250 BLDPBD00892311-100
4-(2-Methylbutan-2-yl)cyclohexan-1-amine hydrochloride 95%
4-(2-Methylbutan-2-yl)cyclohexan-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₁H₂₄ClN
MW: 205,7700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09971351
CAS číslo: 1177354-16-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR