Tisk…

2-Amino-5-fluoro-3-methylbenzoic acid 95%

BLDPBD00892796-1GEA 4110 CZK
BLDPBD00892796-1G BLDPBD00892796-100 BLDPBD00892796-250 BLDPBD00892796-5G
2-Amino-5-fluoro-3-methylbenzoic acid 95%
2-Amino-5-fluoro-3-methylbenzoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₈H₈FNO₂
MW: 169,153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06409107
CAS číslo: 874804-25-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR