Tisk…

1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]piperazine 90%

BLDPBD00911576-100EA 1970 CZK
BLDPBD00911576-100 BLDPBD00911576-250 BLDPBD00911576-1G
1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]piperazine 90%
1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]piperazine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₉H₁₅N₅O₂
MW: 225,248 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16076302
CAS číslo: 1248142-10-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR