Tisk…

3-Ethyloxan-4-one 95%

BLDPBD00922724-100EA 5820 CZK
BLDPBD00922724-100 BLDPBD00922724-50M BLDPBD00922724-1G BLDPBD00922724-250
3-Ethyloxan-4-one 95%
3-Ethyloxan-4-one

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₇H₁₂O₂
MW: 128,1700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18374985
CAS číslo: 21398-42-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR