Tisk…

(Prop-2-yn-1-yl)bis[(pyridin-2-yl)methyl]amine 97%

BLDPBD00924607-1GEA 11030 CZK
BLDPBD00924607-1G BLDPBD00924607-250 BLDPBD00924607-100
(Prop-2-yn-1-yl)bis[(pyridin-2-yl)methyl]amine 97%
(Prop-2-yn-1-yl)bis[(pyridin-2-yl)methyl]amine

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₅H₁₅N₃
MW: 237,3100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30833928
CAS číslo: 950984-75-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR