Tisk…

N-{[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}acetamide 95%

BLDPBD00927571-1GEA 15140 CZK
BLDPBD00927571-1G BLDPBD00927571-250 BLDPBD00927571-100
N-{[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}acetamide 95%
N-{[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}acetamide

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₅H₂₂BNO₃
MW: 275,151 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18383571
CAS číslo: 1235450-93-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR