Tisk…

(S)-N-(4-chlorobenzylidene)-2-methylpropane-2-sulphinamide 97%

BLDPBD00928184-1GEA 4390 CZK
BLDPBD00928184-1G BLDPBD00928184-250 BLDPBD00928184-5G
(S)-N-(4-chlorobenzylidene)-2-methylpropane-2-sulphinamide 97%
(S)-N-(4-chlorobenzylidene)-2-methylpropane-2-sulphinamide

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₁H₁₄ClNOS
MW: 243,753 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18781478
CAS číslo: 851513-47-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR