Tisk…

(2S,5R)-1-tert-Butyl 2-ethyl 5-((benzyloxy)amino)piperidine-1,2-dicarboxylate 98%

BLDPBD00930951-5GEA 8780 CZK
BLDPBD00930951-5G BLDPBD00930951-250 BLDPBD00930951-1G
(2S,5R)-1-tert-Butyl 2-ethyl 5-((benzyloxy)amino)piperidine-1,2-dicarboxylate 98%
(2S,5R)-1-tert-Butyl 2-ethyl 5-((benzyloxy)amino)piperidine-1,2-dicarboxylate

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₂₀H₃₀N₂O₅
MW: 378,4626 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31382304
CAS číslo: 1463501-46-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR