Tisk…

(S)-1-(3-chloro-2-fluorophenyl)ethan-1-amine 95%

BLDPBD00932927-250EA 8730 CZK
BLDPBD00932927-250 BLDPBD00932927-1G BLDPBD00932927-100
(S)-1-(3-chloro-2-fluorophenyl)ethan-1-amine 95%
(S)-1-(3-chloro-2-fluorophenyl)ethan-1-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₈H₉ClFN
MW: 173,6200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07772566
CAS číslo: 1228561-56-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR