Tisk…

(E)-3-(4-Fluorophenyl)prop-2-en-1-ol 97%

BLDPBD00933395-100EA 4480 CZK
BLDPBD00933395-100 BLDPBD00933395-1G BLDPBD00933395-250
(E)-3-(4-Fluorophenyl)prop-2-en-1-ol 97%
(E)-3-(4-Fluorophenyl)prop-2-en-1-ol

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₉FO
MW: 152,1656 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848635
CAS číslo: 124980-95-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR