Tisk…

5,10,15,20-meso-Tetrakis[4-(methoxycarbonyl)phenyl]porphyrin 96%

BLDPBD00934647-10GEA 19160 CZK
BLDPBD00934647-10G BLDPBD00934647-1G BLDPBD00934647-5G
5,10,15,20-meso-Tetrakis[4-(methoxycarbonyl)phenyl]porphyrin 96%
5,10,15,20-meso-Tetrakis[4-(methoxycarbonyl)phenyl]porphyrin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₅₂H₃₈N₄O₈
MW: 846,8801 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00218869
CAS číslo: 22112-83-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR