Tisk…

tert-Butyl 4-[hydroxy(phenyl)methyl]piperidine-1-carboxylate 98%

BLDPBD00935852-100EA 5280 CZK
BLDPBD00935852-100 BLDPBD00935852-1G BLDPBD00935852-250
tert-Butyl 4-[hydroxy(phenyl)methyl]piperidine-1-carboxylate 98%
tert-Butyl 4-[hydroxy(phenyl)methyl]piperidine-1-carboxylate

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₇H₂₅NO₃
MW: 291,3853 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12912653
CAS číslo: 269740-46-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR