Tisk…

Azepane-4-carboxylic acid hydrochloride 95%

BLDPBD00935984-1GEA 11320 CZK
BLDPBD00935984-1G BLDPBD00935984-100 BLDPBD00935984-250
Azepane-4-carboxylic acid hydrochloride 95%
Azepane-4-carboxylic acid hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₁₄ClNO₂
MW: 179,6446 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24387387
CAS číslo: 1393449-23-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR