Tisk…

2,4-Dimethoxy-5-methylaniline 95%

BLDPBD00937383-1GEA 21890 CZK
BLDPBD00937383-1G BLDPBD00937383-100 BLDPBD00937383-250
2,4-Dimethoxy-5-methylaniline 95%
2,4-Dimethoxy-5-methylaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₉H₁₃NO₂
MW: 167,2050 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18431944
CAS číslo: 82477-59-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR