Tisk…

2,2,3,3-Tetramethyl-4,7,10,13-tetraoxa-3-silapentadecan-15-ol 95%

BLDPBD00941471-1GEA 2460 CZK
BLDPBD00941471-1G BLDPBD00941471-250 BLDPBD00941471-5G
2,2,3,3-Tetramethyl-4,7,10,13-tetraoxa-3-silapentadecan-15-ol 95%
2,2,3,3-Tetramethyl-4,7,10,13-tetraoxa-3-silapentadecan-15-ol

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₄H₃₂O₅Si
MW: 308,4864 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD25424114
CAS číslo: 134179-40-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR