Tisk…

(2S,2'S,3S,3'S)-WingPhos 97%

BLDPBD00941749-250EA 10410 CZK
BLDPBD00941749-250 BLDPBD00941749-100 BLDPBD00941749-50M
(2S,2'S,3S,3'S)-WingPhos 97%
(2S,2'S,3S,3'S)-WingPhos

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₅₀H₄₄O₂P₂
MW: 738,831 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31630708
CAS číslo: 1435940-19-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR