Tisk…

3-Cyclohexylidene-1,3-dihydro-2H-indol-2-one 90%

BLDPBD00944496-1GEA 10260 CZK
BLDPBD00944496-1G BLDPBD00944496-250 BLDPBD00944496-100
3-Cyclohexylidene-1,3-dihydro-2H-indol-2-one 90%
3-Cyclohexylidene-1,3-dihydro-2H-indol-2-one

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₁₄H₁₅NO
MW: 213,2750 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00449408
CAS číslo: 3478-78-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR