Tisk…

N,9,9-Triphenyl-9H-fluoren-2-amine 97%

BLDPBD00944952-100EA 456 CZK
BLDPBD00944952-100 BLDPBD00944952-250 BLDPBD00944952-1G
N,9,9-Triphenyl-9H-fluoren-2-amine 97%
N,9,9-Triphenyl-9H-fluoren-2-amine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₃₁H₂₃N
MW: 409,53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30470208
CAS číslo: 860465-14-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR