(S,S)-iPr-BI-DIME 97%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD00946012-100EA
3540
CZK
BLDPBD00946012-100
(S,S)-iPr-BI-DIME 97%
(S,S)-iPr-BI-DIME
Storage condition: Inert atmosphere, 2-8°C
Vzorec:
C₂₂H₂₉O₃P MW: 372,438 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD31630765 CAS číslo: 1477517-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...