Tisk…

[1-(3,4-Dimethoxyphenyl)propyl]amine hydrochloride 95+%

BLDPBD00946181-100EA 3600 CZK
BLDPBD00946181-100 BLDPBD00946181-250
[1-(3,4-Dimethoxyphenyl)propyl]amine hydrochloride 95+%
[1-(3,4-Dimethoxyphenyl)propyl]amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₁H₁₈ClNO₂
MW: 231,7191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18483503
CAS číslo: 104174-57-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR