Tisk…

(4-(Benzyloxy)-2-formyl-5-methoxyphenyl)boronic acid 97%

BLDPBD00946771-100EA 3570 CZK
BLDPBD00946771-100
(4-(Benzyloxy)-2-formyl-5-methoxyphenyl)boronic acid 97%
(4-(Benzyloxy)-2-formyl-5-methoxyphenyl)boronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₅H₁₅BO₅
MW: 286,0900 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27976031
CAS číslo: 1021431-39-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR