Tisk…

(S)-BI-DIME 97%

BLDPBD00946899-250EA 4620 CZK
BLDPBD00946899-250 BLDPBD00946899-1G BLDPBD00946899-100
(S)-BI-DIME 97%
(S)-BI-DIME

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₉H₂₃O₃P
MW: 330,358 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31630893
CAS číslo: 1373432-09-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR