Tisk…

3-(4-Hydroxy-1-oxoisoindolin-2-yl)piperidine-2,6-dione 97%

BLDPBD00947377-250EA 1630 CZK
BLDPBD00947377-250 BLDPBD00947377-1G BLDPBD00947377-5G BLDPBD00947377-25G BLDPBD00947377-100
3-(4-Hydroxy-1-oxoisoindolin-2-yl)piperidine-2,6-dione 97%
3-(4-Hydroxy-1-oxoisoindolin-2-yl)piperidine-2,6-dione

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₃H₁₂N₂O₄
MW: 260,2454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29054753
CAS číslo: 1061604-41-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR