Tisk…

Ethyl 6-isopropyl-2-methyl-4-oxocyclohex-2-ene-1-carboxylate 95%

BLDPBD00948340-1GEA 1320 CZK
BLDPBD00948340-1G BLDPBD00948340-10G BLDPBD00948340-5G
Ethyl 6-isopropyl-2-methyl-4-oxocyclohex-2-ene-1-carboxylate 95%
Ethyl 6-isopropyl-2-methyl-4-oxocyclohex-2-ene-1-carboxylate

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₃H₂₀O₃
MW: 224,2961 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00482052
CAS číslo: 24079-95-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR