Tisk…

2-Bromo-4-fluoro-5-methoxybenzaldehyde 95%

BLDPBD00948541-5GEA 27990 CZK
BLDPBD00948541-5G BLDPBD00948541-250 BLDPBD00948541-1G
2-Bromo-4-fluoro-5-methoxybenzaldehyde 95%
2-Bromo-4-fluoro-5-methoxybenzaldehyde

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₆BrFO₂
MW: 233,034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22371190
CAS číslo: 865186-62-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR