Tisk…

2,3-Dichloro-4-fluorobenzonitrile 95%

BLDPBD00949094-50MEA 998 CZK
BLDPBD00949094-50M BLDPBD00949094-1G BLDPBD00949094-100 BLDPBD00949094-250
2,3-Dichloro-4-fluorobenzonitrile 95%
2,3-Dichloro-4-fluorobenzonitrile

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₇H₂Cl₂FN
MW: 190,0019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19690560
CAS číslo: 908123-82-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR