Tisk…

Pentyl cyclopentanecarboxylate 95%

BLDPBD00949433-250EA 8100 CZK
BLDPBD00949433-250 BLDPBD00949433-1G BLDPBD00949433-100 BLDPBD00949433-5G
Pentyl cyclopentanecarboxylate 95%
Pentyl cyclopentanecarboxylate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₁H₂₀O₂
MW: 184,2753 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32197084
CAS číslo: 959026-72-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR