Tisk…

Methyl 3-bromo-2-phenyl-propanoate 95%

BLDPBD00949739-1GEA 13940 CZK
BLDPBD00949739-1G BLDPBD00949739-250 BLDPBD00949739-100
Methyl 3-bromo-2-phenyl-propanoate 95%
Methyl 3-bromo-2-phenyl-propanoate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₁₁BrO₂
MW: 243,097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17215983
CAS číslo: 99070-19-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR