Tisk…

4-Bromo-2-fluoro-N-(4-iodophenyl)-benzenesulphonamide 95%

BLDPBD00950490-100EA 1120 CZK
BLDPBD00950490-100 BLDPBD00950490-250 BLDPBD00950490-1G
4-Bromo-2-fluoro-N-(4-iodophenyl)-benzenesulphonamide 95%
4-Bromo-2-fluoro-N-(4-iodophenyl)-benzenesulphonamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₂H₈BrFINO₂S
MW: 456,0700 g/mol
Storage Temperature: Ambient
CAS číslo: 1771023-28-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR