Tisk…

Methyl (R)-2-amino-2-(2-bromo-4-fluorophenyl)acetate hcl 95%

BLDPBD00951197-50MEA 3880 CZK
BLDPBD00951197-50M BLDPBD00951197-100 BLDPBD00951197-1G BLDPBD00951197-250
Methyl (R)-2-amino-2-(2-bromo-4-fluorophenyl)acetate hcl 95%
Methyl (R)-2-amino-2-(2-bromo-4-fluorophenyl)acetate hcl

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₉H₁₀BrClFNO₂
MW: 298,5400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12911244
CAS číslo: 1391381-36-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR