Tisk…

2-(Benzyloxy)-1-methylpyridin-1-ium trifluoromethanesulphonate 96%

BLDPBD00953100-5GEA 6730 CZK
BLDPBD00953100-5G BLDPBD00953100-250 BLDPBD00953100-1G
2-(Benzyloxy)-1-methylpyridin-1-ium trifluoromethanesulphonate 96%
2-(Benzyloxy)-1-methylpyridin-1-ium trifluoromethanesulphonate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₁₄H₁₄F₃NO₄S
MW: 349,3255 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09265129
CAS číslo: 882980-43-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR