Tisk…

(S)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-2H-[1,4]oxazino[2,3,4-ij]quinolin-7(3H)-one 95%

BLDPBD00955003-250EA 12490 CZK
BLDPBD00955003-250 BLDPBD00955003-50M BLDPBD00955003-100 BLDPBD00955003-1G
(S)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-2H-[1,4]oxazino[2,3,4-ij]quinolin-7(3H)-one 95%
(S)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-2H-[1,4]oxazino[2,3,4-ij]quinolin-7(3H)-one

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₇H₂₀FN₃O₂
MW: 317,3580 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28016445
CAS číslo: 178964-53-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR