Tisk…

N,N-Bis(9,9-dimethyl-9H-fluoren-2-yl)aniline 95%

BLDPBD00955092-5GEA 5960 CZK
BLDPBD00955092-5G BLDPBD00955092-250 BLDPBD00955092-1G
N,N-Bis(9,9-dimethyl-9H-fluoren-2-yl)aniline 95%
N,N-Bis(9,9-dimethyl-9H-fluoren-2-yl)aniline

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₃₆H₃₁N
MW: 477,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29089346
CAS číslo: 165320-27-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR