Tisk…

(R)-1-(5-(Trifluoromethyl)pyridin-2-yl)ethanamine 95%

BLDPBD00955260-250EA 12970 CZK
BLDPBD00955260-250 BLDPBD00955260-100
(R)-1-(5-(Trifluoromethyl)pyridin-2-yl)ethanamine 95%
(R)-1-(5-(Trifluoromethyl)pyridin-2-yl)ethanamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₈H₉F₃N₂
MW: 190,1657 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09824132
CAS číslo: 1213052-96-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR