Tisk…

7-Methoxy-3-propyl-1H-pyrrolo[2,3-c]pyridine 97%

BLDPBD00955760-250EA 14080 CZK
BLDPBD00955760-250 BLDPBD00955760-100 BLDPBD00955760-1G
7-Methoxy-3-propyl-1H-pyrrolo[2,3-c]pyridine 97%
7-Methoxy-3-propyl-1H-pyrrolo[2,3-c]pyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₁₁H₁₄N₂O
MW: 190,2417 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28674132
CAS číslo: 1493855-75-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR