Tisk…

Cyclopropyl(4,6-dichloropyridin-3-yl)methanone 95%

BLDPBD00957150-250EA 12430 CZK
BLDPBD00957150-250 BLDPBD00957150-1G BLDPBD00957150-100
Cyclopropyl(4,6-dichloropyridin-3-yl)methanone 95%
Cyclopropyl(4,6-dichloropyridin-3-yl)methanone

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₇Cl₂NO
MW: 216,0640 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31543872
CAS číslo: 1863098-74-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR