Tisk…

(S)-6,6'-Dibromo-[1,1'-binaphthalene]-2,2'-diamine 98%

BLDPBD00957165-100EA 6080 CZK
BLDPBD00957165-100 BLDPBD00957165-1G BLDPBD00957165-250
(S)-6,6'-Dibromo-[1,1'-binaphthalene]-2,2'-diamine 98%
(S)-6,6'-Dibromo-[1,1'-binaphthalene]-2,2'-diamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₂₀H₁₄Br₂N₂
MW: 442,1466 g/mol
Storage Temperature: Ambient
CAS číslo: 947337-17-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR