Tisk…

(2R,6S)-rel-1,2,6-Trimethylpiperazine 95%

BLDPBD00957516-1GEA 2880 CZK
BLDPBD00957516-1G BLDPBD00957516-250
(2R,6S)-rel-1,2,6-Trimethylpiperazine 95%
(2R,6S)-rel-1,2,6-Trimethylpiperazine
Vzorec: C₇H₁₆N₂
MW: 128,22 g/mol
MDL Number: MFCD19217030
CAS číslo: 147539-61-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR