Tisk…

(S)-3-(3-chlorophenyl)-3-hydroxypropanoic acid 97%

BLDPBD00958539-250EA 2770 CZK
BLDPBD00958539-250 BLDPBD00958539-5G BLDPBD00958539-100 BLDPBD00958539-1G
(S)-3-(3-chlorophenyl)-3-hydroxypropanoic acid 97%
(S)-3-(3-chlorophenyl)-3-hydroxypropanoic acid

Storage condition: Sealed in dry, Store in freezer, under -20°C

Vzorec: C₉H₉ClO₃
MW: 200,6190 g/mol
Storage Temperature: Freezer
MDL Number: MFCD24479954
CAS číslo: 625095-56-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR