Tisk…

(11bR)-N-(2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulphonamide 97%

BLDPBD00960558-250EA 11750 CZK
BLDPBD00960558-250 BLDPBD00960558-1G BLDPBD00960558-100
(11bR)-N-(2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulphonamide 97%
(11bR)-N-(2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulphonamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₃₇H₁₇F₁₅NO₅PS
MW: 903,5494 g/mol
Storage Temperature: Ambient
CAS číslo: 1010800-02-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR