Tisk…

3,5-Di(1H-imidazol-1-yl)pyridine 97%

BLDPBD00961418-1GEA 9500 CZK
BLDPBD00961418-1G BLDPBD00961418-100 BLDPBD00961418-250
3,5-Di(1H-imidazol-1-yl)pyridine 97%
3,5-Di(1H-imidazol-1-yl)pyridine

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₁H₉N₅
MW: 211,2227 g/mol
Storage Temperature: Ambient
CAS číslo: 1374155-84-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR