Tisk…

(R)-4-(Benzyloxy)-3-(((benzyloxy)carbonyl)amino)-4-oxobutanoic acid 97%

BLDPBD00961589-1GEA 285 CZK
BLDPBD00961589-1G BLDPBD00961589-5G
(R)-4-(Benzyloxy)-3-(((benzyloxy)carbonyl)amino)-4-oxobutanoic acid 97%
(R)-4-(Benzyloxy)-3-(((benzyloxy)carbonyl)amino)-4-oxobutanoic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₉H₁₉NO₆
MW: 357,3573 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00672532
CAS číslo: 81440-35-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR