Tisk…

(2S,2'S)-[N,N'-Bis(2-pyridylmethyl)]-2,2'-bipyrrolidine tetrahydrochloride 98%

BLDPBD00961917-5GEA 50220 CZK
BLDPBD00961917-5G BLDPBD00961917-100 BLDPBD00961917-250 BLDPBD00961917-1G
(2S,2'S)-[N,N'-Bis(2-pyridylmethyl)]-2,2'-bipyrrolidine tetrahydrochloride 98%
(2S,2'S)-[N,N'-Bis(2-pyridylmethyl)]-2,2'-bipyrrolidine tetrahydrochloride
Vzorec: C₂₀H₃₀Cl₄N₄
MW: 468,29 g/mol
MDL Number: MFCD16621417
CAS číslo: 1285550-13-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR