Tisk…

(S)-2-(4-(Piperidin-3-yl)phenyl)-2H-indazole-7-carboxamide 4-methylbenzenesulphonate hydrate 98%

BLDPBD00961981-1GEA 5050 CZK
BLDPBD00961981-1G BLDPBD00961981-250 BLDPBD00961981-100
(S)-2-(4-(Piperidin-3-yl)phenyl)-2H-indazole-7-carboxamide 4-methylbenzenesulphonate hydrate 98%
(S)-2-(4-(Piperidin-3-yl)phenyl)-2H-indazole-7-carboxamide 4-methylbenzenesulphonate hydrate

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₂₆H₃₀N₄O₅S
MW: 510,6052 g/mol
Storage Temperature: Ambient
CAS číslo: 1613220-15-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR