Tisk…

1-[4-(Dimethylamino)-2-hydroxyphenyl]ethan-1-one 95%

BLDPBD00962231-250EA 2090 CZK
BLDPBD00962231-250 BLDPBD00962231-5G BLDPBD00962231-100 BLDPBD00962231-1G
1-[4-(Dimethylamino)-2-hydroxyphenyl]ethan-1-one 95%
1-[4-(Dimethylamino)-2-hydroxyphenyl]ethan-1-one

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₁₃NO₂
MW: 179,2157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11870193
CAS číslo: 107070-69-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR