Tisk…

3-Bromo-2,5-dimethylthiophene 95%

BLDPBD00962420-100EA 1660 CZK
BLDPBD00962420-100 BLDPBD00962420-1G BLDPBD00962420-5G BLDPBD00962420-250
3-Bromo-2,5-dimethylthiophene 95%
3-Bromo-2,5-dimethylthiophene

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₆H₇BrS
MW: 191,088 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18451735
CAS číslo: 31819-37-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR