Tisk…

2-Bromo-4-methyl-1-nitrobenzene 95%

BLDPBD00963069-1GEA 884 CZK
BLDPBD00963069-1G BLDPBD00963069-5G
2-Bromo-4-methyl-1-nitrobenzene 95%
2-Bromo-4-methyl-1-nitrobenzene
Vzorec: C₇H₆BrNO₂
MW: 216,03 g/mol
MDL Number: MFCD07363742
CAS číslo: 40385-54-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR