Tisk…

(R)-BI-DIME 97%

BLDPBD00965637-250EA 4450 CZK
BLDPBD00965637-250 BLDPBD00965637-1G BLDPBD00965637-100
(R)-BI-DIME 97%
(R)-BI-DIME

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₉H₂₃O₃P
MW: 330,358 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31630704
CAS číslo: 1338454-03-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR