Tisk…

2-Ethyl-1,1'-biphenyl 98%

BLDPBD00968641-1GEA 2170 CZK
BLDPBD00968641-1G BLDPBD00968641-250 BLDPBD00968641-5G
2-Ethyl-1,1'-biphenyl 98%
2-Ethyl-1,1'-biphenyl

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₄H₁₄
MW: 182,2610 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30296301
CAS číslo: 1812-51-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR