Tisk…

Ethylene Glycol 1,2-Bis(2-propynyl) ether 98%

BLDPBD00968801-250EA 399 CZK
BLDPBD00968801-250 BLDPBD00968801-5G BLDPBD00968801-1G
Ethylene Glycol 1,2-Bis(2-propynyl) ether 98%
Ethylene Glycol 1,2-Bis(2-propynyl) ether

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₁₀O₂
MW: 138,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03640302
CAS číslo: 40842-04-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR